Order Entry
Germany
ContactUsLinkComponent
Anti-Ly-6G (Gr-1) Rat Antibody (redFluor™ 710) [clone: RB6-8C5]
Anti-Ly-6G (Gr-1) Rat Antibody (redFluor™ 710) [clone: RB6-8C5]
Supplier:  Tonbo Biosciences
Certificates
Anti-Ly-6G (Gr-1) Rat Antibody (redFluor™ 710) [clone: RB6-8C5]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 80-5931-U100
  • 80-5931-U025
  • Antigen symbol:
  • LY6C1
  • LY6C1
  • Antigen name:
  • Lymphocyte Antigen 6 Complex, locus C1
  • Lymphocyte Antigen 6 Complex, locus C1
  • Antigen synonyms:
  • Ly6c|AA959465|Ly-6C|Ly-6C1|AA682074
  • Ly6c|AA959465|Ly-6C|Ly-6C1|AA682074
  • Antibody type:
  • Primary
  • Primary
  • Conjugation:
  • redFluor® 710
  • redFluor® 710
  • Clone:
  • RB6-8C5
  • RB6-8C5
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 17067
  • 17067
  • Isotype:
  • IgG2b
  • IgG2b
  • Flow cytometry:
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Storage temperature:
  • Store undiluted at 2-8 °C, protect from light, do not freeze.
  • Store undiluted at 2-8 °C, protect from light, do not freeze.
  • Tested applications:
  • Flow cytometry
  • Flow cytometry

Specifications

Related Information