Order Entry
Germany
ContactUsLinkComponent
Mouse IgG1 Isotype Control [clone: MOPC-21]
Mouse IgG1 Isotype Control [clone: MOPC-21]
Catalog # 70-4714-U100
Supplier:  Tonbo Biosciences
Mouse IgG1 Isotype Control [clone: MOPC-21]
Catalog # 70-4714-U100
Supplier:  Tonbo Biosciences

Specifications

  • Antibody type:
    Control
  • Clone:
    MOPC-21
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Storage temperature:
    Store Undiluted at 2 to 8 °C
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

The MOPC-21 immunoglobulin is useful as an isotype-matched control. The MOPC-21 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG1 antibodies.

Type: Control
Antigen: MOPC-21
Clonality:
Clone: MOPC-21
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: N/A