Nitrile gloves, Solo 999
Supplier: MAPA
Certificates
About this item
These disposable nitrile gloves provide excellent mechanical protection. They are ideal for use in oily environments.
- Excellent dexterity thanks to reduced material thickness
- Chlorinated interior: Easy to put on and to take off
- Smooth with pebbled fingertips for improved grip
- Recommended for people sensitised to natural rubber proteins
Typical applications include: Assembly of small oily parts, work with composite materials (resins), laboratory tasks, manufacturing of medicines, pharmaceutical preparation, research, analysis and the handling of precision parts.
Certifications: EN ISO 374 Type B (JKT), EN ISO 374-5 VIRUS; Cat. III.
Specifications
- Catalog No:
- 112-5615
- 112-5616
- 112-5617
- 112-5618
- Model:
- Solo 999
- Solo 999
- Solo 999
- Solo 999
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Finish:
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- EN norm:
- EN ISO 374 Type B (JKT), EN ISO 374-5 VIRUS; Cat. III
- EN ISO 374 Type B (JKT), EN ISO 374-5 VIRUS; Cat. III
- EN ISO 374 Type B (JKT), EN ISO 374-5 VIRUS; Cat. III
- EN ISO 374 Type B (JKT), EN ISO 374-5 VIRUS; Cat. III