Orders LinkContactUsLinkComponent
Laboratory marker
Laboratory marker
Laboratory marker, Superior, Black
Catalog # MEDT47-1070-00
Supplier:   MEDITE
Laboratory marker
Catalog # MEDT47-1070-00
Supplier:  MEDITE
Laboratory marker
Laboratory marker, Superior, Black
Price
53,40 €
Log in for availability
1 * 12 ST
Product Details & Documents
Marker for labelling cassettes and microscope slides with a high contrast black ink. These are water-based, therefor no early drying out is possible.
  • Water resistant, odour-free and non toxic
  • Resistant against most laboratory chemicals
  • No pretreatment of surfaces necessary
Specifications
  • Colour:
    Black
  • Description:
    Laboratory marker, Superior