Anti-Helios Armenian Hamster Monoclonal Antibody (PB (Pacific Blue)) [clone: 22F6]
Supplier: BIOLEGEND INC
Certificates
About this item
Type: Primary
Antigen: HELIOS
Clonality: Monoclonal
Clone:
Conjugation: PB (Pacific Blue)
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse
Specifications
- Catalog No:
- BLEG137220
- BLEG137210
- Antigen symbol:
- IKZF2
- IKZF2
- Antigen name:
- Helios
- Helios
- Antigen synonyms:
- IKAROS family zinc finger 2|ZNF1A2|ANF1A2|ZNFN1A2
- IKAROS family zinc finger 2|ZNF1A2|ANF1A2|ZNFN1A2
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PB (Pacific Blue)
- PB (Pacific Blue)
- Clone:
- 22F6
- Reactivity:
- Mouse
- Mouse, Cat, Baboon, Capuchin monkey, Pig, Rabbit, Sheep, Cynomolgus monkey, Macaque, Chimpanzee, Human, Bovine, Dog
- Host:
- Armenian hamster
- Armenian hamster
- Gene ID:
- 22779, 22807
- Isotype:
- IgG
- IgG
- Flow cytometry:
- Yes
- Immunogen:
- Helios peptide (aa 51-107)
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Purification:
- The antibody was purified by affinity chromatography, and conjugated with Pacific Blue® under optimal conditions. The solution is free of unconjugated Pacific Blue®.
- Storage temperature:
- The antibody solution should be stored undiluted between 2°C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:
- Ambient RT