Anti-IL18R alpha Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate))
Catalog # BLEG313810
Supplier: BIOLEGEND INC
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 18 Receptor Alpha
- Antigen symbol:IL18R alpha
- Clonality:Monoclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Mouse
- Antigen synonyms:CD218a
- Size:100 Tests
- Pk:100 Tests
Specifications
About this item
Type: Primary
Antigen: IL18R alpha
Clonality: Monoclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human