Anti-CYP1A Mouse Monoclonal Antibody [clone: NP-7]
Supplier: Cayman Chemical
Certificates
About this item
Type: Primary
Antigen: CYP1A
Clonality: Monoclonal
Clone: NP-7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Carp, Cod, Salmon, some flatfish
Specifications
- Catalog No:
- CAYM173125-500
- CAYM173125-100
- Antigen symbol:
- CYP1A
- CYP1A
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- NP-7
- NP-7
- Reactivity:
- Carp, Cod, Flatfish, Salmon
- Carp, Cod, Flatfish, Salmon
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Atlantic cod (Gadus morhua L.) CYP1A, from liver microsomes of -betanaphthoflavone-treated cod
- Atlantic cod (Gadus morhua L.) CYP1A, from liver microsomes of -betanaphthoflavone-treated cod
- Purification:
- purified
- purified
- Cross adsorption:
- No
- No
- Storage buffer:
- 0,1 M sodium phosphate, pH 7,4, containing 0,15 M sodium chloride, 12.3 mM sodium azide, and 1% BSA
- 0,1 M sodium phosphate, pH 7,4, containing 0,15 M sodium chloride, 12.3 mM sodium azide, and 1% BSA
- Storage temperature:
- +4 °C
- +4 °C
- Shipping temperature:
- 4 °C
- 4 °C