Anti-Vtg Mouse Monoclonal Antibody [clone: KB-1]
Supplier: Cayman Chemical
Certificates
About this item
Type: Primary
Antigen: Vtg
Clonality: Monoclonal
Clone: KB-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Atlantic Salmon, Brown Trout, Salmonella
Specifications
- Catalog No:
- CAYM170147-500
- CAYM170147-100
- Antigen symbol:
- VTG
- VTG
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- KB-1
- KB-1
- Reactivity:
- Salmonella, Brown trout, Atlantic salmon
- Salmonella, Brown trout, Atlantic salmon
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Vtg from plasma of 17-beta estradiol treated Atlantic salmon (S. salar)
- Vtg from plasma of 17-beta estradiol treated Atlantic salmon (S. salar)
- Purification:
- purified
- purified
- Cross adsorption:
- No
- No
- Storage buffer:
- 0,1 M sodium phosphate, pH 7,4, containing 0,15 M sodium chloride, 12.3 mM sodium azide, and 1% BSA
- 0,1 M sodium phosphate, pH 7,4, containing 0,15 M sodium chloride, 12.3 mM sodium azide, and 1% BSA
- Storage temperature:
- +4 °C
- +4 °C
- Shipping temperature:
- 4 °C
- 4 °C