Anti-Vtg Rabbit Polyclonal Antibody [clone: PO-1]
Supplier: Cayman Chemical
Certificates
About this item
Type: Primary
Antigen: Vtg
Clonality: Polyclonal
Clone: PO-1
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Arctic Char, Atlantic Cod, Atlantic Salmon, brown Trout, Flounder, Tilapia, Turbot, Wrasse, Zebrafish
Specifications
- Catalog No:
- CAYM170120-500
- CAYM170120-100
- Antigen symbol:
- VTG
- VTG
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- PO-1
- PO-1
- Reactivity:
- Zebrafish, Arctic char, Atlantic cod, Flounder, Tilapia, Turbot, Brown trout, Wrasse, Atlantic salmon
- Zebrafish, Arctic char, Atlantic cod, Flounder, Tilapia, Turbot, Brown trout, Wrasse, Atlantic salmon
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Vtg from plasma of 17-beta-estradiol treated Arctic char (Salvelinus alpinus)
- Vtg from plasma of 17-beta-estradiol treated Arctic char (Salvelinus alpinus)
- Purification:
- purified
- purified
- Cross adsorption:
- No
- No
- Storage buffer:
- 0,1 M sodium phosphate, pH 7,4, containing 0,15 M sodium chloride, 12.3 mM sodium azide, and 1% BSA
- 0,1 M sodium phosphate, pH 7,4, containing 0,15 M sodium chloride, 12.3 mM sodium azide, and 1% BSA
- Storage temperature:
- +4 °C
- +4 °C
- Shipping temperature:
- 4 °C
- 4 °C