Anti-Vtg Mouse Monoclonal Antibody [clone: ND-5C9]
Supplier: Cayman Chemical
Certificates
About this item
Type: Primary
Antigen: Vtg
Clonality: Monoclonal
Clone: ND-5C9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Flounder, Halibut, Sheepshead Minnow
Specifications
- Catalog No:
- CAYM170100-500
- CAYM170100-100
- Antigen symbol:
- VTG
- VTG
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- ND-5C9
- ND-5C9
- Reactivity:
- Sheepshead minnow, Flounder, Halibut
- Sheepshead minnow, Flounder, Halibut
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Vtg from sheepshead minnow (C. variegatus), bowfin (A. calva), brown bullhead (A. nebulosus), and spotted gar (L. oculatus)
- Vtg from sheepshead minnow (C. variegatus), bowfin (A. calva), brown bullhead (A. nebulosus), and spotted gar (L. oculatus)
- Purification:
- purified by affinity chromatography
- purified by affinity chromatography
- Cross adsorption:
- No
- No
- Storage buffer:
- 0,1 M sodium phosphate, pH 7,4, containing 0,15 M sodium chloride, 1% BSA, and 12.3 M sodium azide
- 0,1 M sodium phosphate, pH 7,4, containing 0,15 M sodium chloride, 1% BSA, and 12.3 M sodium azide
- Storage temperature:
- +4 °C
- +4 °C
- Shipping temperature:
- 4 °C
- 4 °C