Order Entry
Germany
ContactUsLinkComponent
Anti-BLT1 Mouse Monoclonal Antibody [clone: 7B1]
Anti-BLT1 Mouse Monoclonal Antibody [clone: 7B1]
Catalog # CAYM120111-1
Supplier:  Cayman Chemical
Anti-BLT1 Mouse Monoclonal Antibody [clone: 7B1]
Catalog # CAYM120111-1
Supplier:  Cayman Chemical

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    BLT1
  • Clonality:
    Monoclonal
  • Clone:
    7B1
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Storage buffer:
    TBS, pH 7,4
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    HeLa cells transfected with human BLT1
  • Purification:
    purified by Protein A chromatography
  • Size:
    1 Ea
  • Pk:
    1

Specifications

About this item

Type: Primary
Antigen: BLT1
Clonality: Monoclonal
Clone: 7B1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human