Order Entry
Germany
ContactUsLinkComponent
Anti-LAMP2 Rat Monoclonal Antibody [clone: GL2A7]
Anti-LAMP2 Rat Monoclonal Antibody [clone: GL2A7]
Supplier:  Cayman Chemical
Certificates
Anti-LAMP2 Rat Monoclonal Antibody [clone: GL2A7]
Supplier:  Cayman Chemical

Specifications

  • Catalog No:
  • CAYM10011444-25
  • CAYM10011444-100
  • Antigen symbol:
  • LAMP2
  • LAMP2
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • GL2A7
  • GL2A7
  • Reactivity:
  • Rabbit, Mouse
  • Rabbit, Mouse
  • Host:
  • Rat
  • Rat
  • Isotype:
  • IgG
  • IgG
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • Immunogen:
  • preparation of mouse liver lysosomal membranes
  • preparation of mouse liver lysosomal membranes
  • Purification:
  • purified by Protein G chromatography
  • purified by Protein G chromatography
  • Cross adsorption:
  • No
  • No
  • Storage buffer:
  • TBS, containing 0,09% sodium azide and 50% glycerol
  • TBS, containing 0,09% sodium azide and 50% glycerol
  • Storage temperature:
  • –20 °C
  • –20 °C
  • Shipping temperature:
  • –20 °C
  • –20 °C

Specifications