Anti-PGR Mouse Monoclonal Antibody [clone: 1154]
CAYM10009762-1
- Antibody type:Primary
- Antigen symbol:PGR
- Clonality:Monoclonal
- Clone:1154
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Modification:phospho-Ser294
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:phosphopeptide corresponding to amino acid residues surrounding the phospho-Ser294 of human progesterone receptor
- Purification:purified by affinity chromatography
- Size:1 Ea
- Pk:1
Type: Primary
Antigen: PGR
Clonality: Monoclonal
Clone: 1154
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human