Anti-Nitrotyrosine Mouse Monoclonal Antibody (Biotin) [clone: 8C7.3]
Catalog # CAYM10006966-100
Supplier: Cayman Chemical
Specifications
- Antibody type:Primary
- Antigen name:Nitrotyrosine
- Clonality:Monoclonal
- Clone:8C7.3
- Conjugation:Biotin
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Multiple
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:peroxynitrite-treated KLH
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Nitrotyrosine
Clonality: Monoclonal
Clone: 8C7.3
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: species independent