Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # CAYM10006588-1
Supplier: Cayman Chemical
Specifications
- Antibody type:Secondary
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Goat
- Reactivity:Rabbit
- Cross adsorption:No
- Storage buffer:PBS, pH 7,2, containing 50% glycerol, 11.3 mg/ml BSA, and 0,04% sodium azide
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Size:1 Ea
- Pk:1
Specifications
About this item
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype:
Reactivity: Rabbit