Anti-CD8 Mouse Monoclonal Antibody [clone: RIV11]
CAYM10004715-1
- Antibody type:Primary
- Antigen symbol:CD8
- Clonality:Monoclonal
- Clone:RIV11
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Cross adsorption:No
- Storage buffer:PBS, pH 7,2
- Storage temperature:+4 °C
- Shipping temperature:4 °C
- Purification:purified by Protein A chromatography
- Size:1 Ea
- Pk:1
Type: Primary
Antigen: CD8
Clonality: Monoclonal
Clone: RIV11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human