Order Entry
Germany
ContactUsLinkComponent
Anti-BACH2 Rabbit Polyclonal Antibody
Anti-BACH2 Rabbit Polyclonal Antibody
Catalog # ROCK600-401-H35
undefined
Anti-BACH2 Rabbit Polyclonal Antibody
Catalog # ROCK600-401-H35

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    BTB and CNC homology 2
  • Antigen symbol:
    BACH2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Cross adsorption:
    No
  • Format:
    Antibody fraction
  • Form:
    Liquid (sterile filtered)
  • Gene ID:
    12014
  • Antigen synonyms:
    E030004N02Rik
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Dry Ice
  • Immunogen:
    Anti-Bach2 antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to the n-terminus region of the Bach2 Protein.
  • Purification:
    affinity-purified
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Affinity purified Bach-2 antibody has been tested for use in ELISA, Immunoprecipitation, and western blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 91-140 kDa in size corresponding to Bach2 protein.

Recommended Dilutions: ELISA: 1:5000; Western Blot: 1 µg/ml; Immunoprecipitation: 1 µg/ml;

Type: Primary
Antigen: BACH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse