Order Entry
Germany
ContactUsLinkComponent
Anti-GRIN1 Rabbit Polyclonal Antibody
Anti-GRIN1 Rabbit Polyclonal Antibody
Catalog # ROCK600-401-D86
undefined
Anti-GRIN1 Rabbit Polyclonal Antibody
Catalog # ROCK600-401-D86

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Glutamate receptor, ionotropic, N-methyl D-aspartate 1
  • Antigen symbol:
    GRIN1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Antibody fraction
  • Form:
    Lyophilized
  • Gene ID:
    24408
  • Antigen synonyms:
    NMDAR1|GluN1|NR1
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ambient
  • Immunogen:
    Anti-NMDA R1, Splice Variant C2 Antibody was produced by repeated immunizations with a synthetic peptide corresponding to amino acid residues specific to the NR1 subunit, C2 splice variant insert.
  • Purification:
    affinity-purified
  • Size:
    25 µg
  • Pk:
    25 µG

Specifications

About this item

Anti-NMDA R1, Splice Variant C2 (Rabbit) Antibody is suitable for use in Western Blotting and IHC. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 120 kDa in size corresponding to the NMDA Receptor.

Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:1000-2000;
0.05 M Tris Chloride, 0.15 M Sodium Chloride, pH 7.4;

Type: Primary
Antigen: GRIN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat