Order Entry
Germany
ContactUsLinkComponent
Anti-GRIN2B Rabbit Polyclonal Antibody
Anti-GRIN2B Rabbit Polyclonal Antibody
Catalog # ROCK100-401-C87
undefined
Anti-GRIN2B Rabbit Polyclonal Antibody
Catalog # ROCK100-401-C87

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Glutamate receptor, ionotropic, N-methyl D-aspartate 2B
  • Antigen symbol:
    GRIN2B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Cross adsorption:
    No
  • Format:
    Antiserum
  • Form:
    Liquid
  • Gene ID:
    24410
  • Antigen synonyms:
    GluN2B
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Dry Ice
  • Immunogen:
    Anti-NMDA R2B Antibody was produced by repeated immunizations with a fusion Protein from the C-terminal region of the NR2B subunit.
  • Size:
    50 µl
  • Pk:
    50 µl

Specifications

About this item

Anti-NMDA R2B Antibody is suitable for use in Western Blotting. Expect a band of approximately 180kDa in size corresponding to the NR2B subunit of the NMDA receptor in appropriate cell lysate or extract. A dilution of 1:1000 is recommended in western blot.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: GRIN2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Mouse, Human, Rat