Order Entry
Germany
ContactUsLinkComponent
 
Anti-SSA-60/Ro Rabbit Polyclonal Antibody
  USBIS6600-75-100
Anti-SSA-60/Ro Rabbit Polyclonal Antibody
  USBIS6600-75-100

 

  • Antibody type:
    Primary
  • Antigen symbol:
    SSA-60
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Serum
  • Form:
    liquid
  • Antigen synonyms:
    RO
  • Storage buffer:
    PBS, 0,05% sodlium azide, 40% glycerol.
  • Storage temperature:
    –20 °C
  • Immunogen:
    Synthetic peptide corresponding to human SSA-60
  • Size:
    100 μl
  • Pk:
    100 µl

 

 

Recommended dilution: Western Blot: 1:500-1:5000

Type: Primary
Antigen: SSA-60/Ro
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,