Anti-NaDC-1 Rabbit Polyclonal Antibody
USBIN0015B-100
- Antibody type:Primary
- Antigen symbol:NADC-1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,1% BSA.
- Storage temperature:–20 °C
- Concentration:~1 mg/ml
- Immunogen:Synthetic peptide corresponding to 15aa near the C-terminal of mouse NaDC-1 (KLH).
- Purification:purified by immunoaffinity chromatography
- Size:100 μg
- Pk:100 µG
Recommended dilution: Western Blot: 1-10 ?g/ml; ELISA: 0.5-1 ?g/ml
Type: Primary
Antigen: NaDC-1
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse,