Anti-PsaC Rabbit Polyclonal Antibody
Catalog # AGRIAS10939
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:PSAC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:C. reinhardtii,Synechococcus sp. pcc 7942,A. thaliana,N. tabacum,H. vulgare
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide conserved in all known PsaC proteins includingArabidopsis thaliana AtCg01060, Hordeum vulgare P69416, Oryza sativa P0C360, Chlamydomonas reinhardtii Q00914, Synechococcus elongatus Q31QV2
- Size:200 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: PSAC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, C. reinhardtii, H. vulgare, N. tabacum, Synechococcus sp. pcc7942