Order Entry
Germany
ContactUsLinkComponent
Anti-IgY Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Anti-IgY Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Catalog # AGRIAS101104
Supplier:  Agrisera
Anti-IgY Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Catalog # AGRIAS101104
Supplier:  Agrisera

Specifications

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgY
  • Clonality:
    Polyclonal
  • Conjugation:
    TRITC (Tetramethylrhodamine Isothiocyanate)
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Chicken
  • Cross adsorption:
    No
  • Storage temperature:
    +2°C to +8 °C
  • Shipping temperature:
    Ambient
  • Immunogen:
    Purified Chicken IgY, whole molecule
  • Purification:
    purified
  • Size:
    2 mg
  • Pk:
    2 mg

Specifications

About this item

Type: Secondary
Antigen: IgY
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope:
Host: Goat
Isotype:
Reactivity: Chicken