Anti-HSPA5 Rat Monoclonal Antibody [clone: 76-E6]
Catalog # ORIGTA328008
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:HSPA5
- Clonality:Monoclonal
- Clone:76-E6
- Conjugation:Unconjugated
- Host:Rat
- Isotype:IgG2a
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Storage buffer:Phosphate-buffered solution, pH 7,2, containing 0,09% sodium azide.
- Molecular weight:78 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Mouse BiP:u chain complexes
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: HSPA5
Clonality: Monoclonal
Clone: 76-E6
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse