Anti-FOXO1 Rabbit Polyclonal Antibody
Catalog # ORIGTA325474
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:FOXO1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Modification:phospho-Ser256
- Storage buffer:Rabbit IgG in phosphate buffered saline , pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Molecular weight:69 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:The antiserum was produced against A synthesized peptide derived from human FKHR around the phosphorylation site of Serine 256
- Purification:purified by affinity chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200
Type: Primary
Antigen: FOXO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat