

Ethyl acetate ≥99.9%, PESTINORM® Supra Trace for organic trace analysis
Catalog # 85387.320
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Assay (on anhydrous substance):Min. 99.9 %
- Evaporation residue:Max 0.0003%
- Colouration:Max 10 APHA
- Water:Max 0.03%
- GC/ECD Dioxins, Furans & PCB's:Max 5 ng/l
- GC/ECD any pesticide (as Lindane):Max 5 ng/l
- GC/NPD any pesticide (as Parathion):Max 10 ng/l
- GC/ECD 1,2,4-TCB to daca-PCB (as Lindane):Max 5 pg/ml
- GC/ECD DCM to 1,2,4-TCB (as TCM):Max 2 ng/l
- GC/FID C10 to C40 (as n-Decane):Max 2 ng/l
- GC/MSD C10 to C40 (as Decane; 30-600amu):Max 2 ng/l
- Filtered through 0.2um, filled under inert gas:Conforms
Specification Test Results
About this item
CAS Number:141-78-6
:
Formula:CH₃COOC₂H₅
MDL Number:MFCD00009171
:88.11 g/mol
Merck Index:13,03792
Boiling Pt:77.1 °C (1013 hPa)
Melting Pt:–83 °C
Density:0.902 g/cm³ (20 °C)
EINECS:205-500-4
UN:1173
ADR:3,II
:Filtered through a 0.2 µm filter, packaged under inert gas.
Flash Pt:–4 °C
REACH:01-2119475103-46
:GHS02
:GHS07
Storage Temperature:Ambient