Acetone ≥99.9%, PESTINORM® Supra Trace for organic trace analysis
Catalog # 85384.320
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Appearance:Clear colourless liquid
- Identity (IR):Conforms
- Assay (calculated on anhydrous):Min. 99.9 %
- Residue after evaporation:Max. 0.0003 %(m)
- Water:Max. 0.3 %(m)
- Colour:Max. 10 APHA
- GC/ECD dioxins, furans and PCB's:Max. 5 ng/l
- GC/ECD any pesticide (as Lindane):Max. 5 ng/l
- GC/NPD any pesticide (as Parathion):Max. 10 ng/l
- GC/ECD 1,2,4-TCB to deca-PCB(as Lindane):Max. 5 pg/ml
- GC/ECD DCM to 1,2,4-TCB (as TCM):Max. 2.0 ng/ml
- GC/FID C10 to C40 (as n-Decane):Max. 2.0 ng/ml
- GC/MSD C10 to C40 (as Decane; 30-600amu):Max. 2.0 ng/ml
Specification Test Results
About this item
:
:58.08 g/mol
Merck Index:13,00067
Boiling Pt:56.2 °C (1013 hPa)
Melting Pt:–95.4 °C
Density:0.792 g/cm³ (20 °C)
Flash Pt:<–20 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-662-2
UN:1090
ADR:3,II
:Filtered through a 0.2 µm filter, packaged under inert gas.
REACH:01-2119471330-49
:GHS02
:GHS07
CAS Number:67-64-1
Formula:CH₃COCH₃
MDL Number:MFCD00008765