Septa caps
Supplier: EP SCIENTIFIC
About this item
PP, white.
Septa caps fit VOA vials, bottles and jars. The septum is permanently bonded to the cap and processed to meet EPA performance based specifications for volatile organic analysis.
4 Options Available Below
Product Details & Documents
PP, white.
Septa caps fit VOA vials, bottles and jars. The septum is permanently bonded to the cap and processed to meet EPA performance based specifications for volatile organic analysis.
Specifications for Product Family
Specifications
- Catalog No:
- EPSC24-414/WS-3M
- EPSC297-2400UB
- EPSC297-2414
- EPSCS297-5300
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
Specifications
Product Family Options
Product Information
- ThreadAvailabilityPrice
- EPSC24-414/WS-3MOpen top septum cap, bonded 3,2 mm PTFE-lined silicone septaOpen top septum cap, bonded 3,2 mm PTFE-lined silicone septa24-414Log in for availability1 * 1.000 ST1 440,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.EPSC24-414/WS-3MEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - EPSC297-2400UBOpen top septum cap, bonded 1,5 mm, PTFE-lined silicon septaOpen top septum cap, bonded 1,5 mm, PTFE-lined silicon septa24-414Log in for availability1 * 24 ST77,50 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.EPSC297-2400UBEnvironmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - EPSC297-2414Open top cap, without septumSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.EPSC297-2414Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - EPSCS297-5300Open top septum cap, bonded 1,14 mm, PTFE-lined silicon septaOpen top septum cap, bonded 1,14 mm, PTFE-lined silicon septa53-400Log in for availability1 * 24 ST229,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.EPSCS297-5300Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact
Accessories
Product Information
- Catalog #TypeForØAvailabilityPrice
Product Information
- Catalog #TypeForØAvailabilityPrice
Product Information
- Catalog #TypeForØAvailabilityPrice
- EPSC288-0022BPSilicone, PTFE-lined24-214 VOA vials24 mmLog in for availability1 * 2.000 ST - 2 430,00 €
Product Information
- Catalog #TypeForØAvailabilityPrice
- EPSCS24-400-S2Silicone, PTFE-lined24-214 VOA vials24 mmLog in for availability1 * 144 ST - 287,00 €