Order Entry
Germany
ContactUsLinkComponent
Anti-GRIN2B Rabbit Polyclonal Antibody
Anti-GRIN2B Rabbit Polyclonal Antibody
Catalog # BSBTPB9373
Supplier:  Boster Bio
undefined
Anti-GRIN2B Rabbit Polyclonal Antibody
Catalog # BSBTPB9373
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Glutamate receptor, ionotropic, N-methyl D-aspartate 2B
  • Antigen symbol:
    GRIN2B
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Gene ID:
    2904
  • Antigen synonyms:
    hNR3|GluN2B|NR2B|EIEE27|MRD6|NMDAR2B
  • UniProtKB:
    Q13224
  • Storage temperature:
    At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
  • Immunogen:
    E.coli-derived human NMDAR2B recombinant protein (Position: N1076-D1332). Human NMDAR2B shares 97.7% and 97.3% amino acid (aa) sequence identity with mouse and rat NMDAR2B, respectively.
  • Purification:
    Immunogen affinity purified.
  • Size:
    100 µg/vial
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for Glutamate receptor ionotropic, NMDA 2B(GRIN2B) detection. Tested with WB in Human;Mouse;Rat.

The N-methyl-D-aspartate receptor 2B (NMDAR2B), also names as GRIN2B. The sequence of the predicted 1,484-amino acid human protein is 98% and 96% identical to the sequences of the rat and mouse Nmdar2b proteins, respectively. Nmdar2B gene is located on mouse chromosome 6 between Rho and Ly49 centromerically and Glb telomerically. Mapping of the human NMDAR2B receptor subunit gene (GRIN2B) to chromosome 12p12 overexpression of NMDA receptor 2B (NR2B) in the forebrains of transgenic mice leads to enhanced activation of NMDA receptors, facilitating synaptic potentiation in response to stimulation at 10-100 Hz.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.

WB: The detection limit for NMDAR2B is approximately 0.1ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: GRIN2C
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
, Human