Order Entry
Germany
ContactUsLinkComponent
Anti-Connexin 45/GJA7 Rabbit Antibody
Anti-Connexin 45/GJA7 Rabbit Antibody
Catalog # BSBTPA2206
Supplier:  Boster Bio
undefined
Anti-Connexin 45/GJA7 Rabbit Antibody
Catalog # BSBTPA2206
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Connexin 45/GJA7
  • Antigen symbol:
    Cx45
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    near the C-terminus
  • Cross adsorption:
    No
  • Form:
    Lyophilised
  • Antigen synonyms:
    GJA7
  • Amino acid number:
    308 - 322
  • Sequence:
    KIAYKQNKANTAQEQ
  • Storage temperature:
    –20 °C
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human Connexin 45 (308-322aa KIAYKQNKANTAQEQ), different from the related mouse and rat sequences by one amino acid.
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for Gap junction gamma-1 protein(GJC1) detection. Tested with WB in Human, Mouse, Rat.

Reconstitute with distilled water

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: Connexin 45/GJA7
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat