Extender tools
Supplier: UPCHURCH SCIENTIFIC
Jump to:
About this item
These tools made from aluminium or Delrin® can be used to tighten most knurled nuts in hard to reach places. For precise tightening, the extender tools listed with ¼" hex drives are designed to adapt to any torque wrench with a female ¼" socket.
9 Options Available Below
Product Details & Documents
These tools made from aluminium or Delrin® can be used to tighten most knurled nuts in hard to reach places. For precise tightening, the extender tools listed with ¼" hex drives are designed to adapt to any torque wrench with a female ¼" socket.
Product Family Options
Product Information
- MaterialUse with head styleAvailabilityPrice
- UPCHP-291Extender tool, with 1/4"hex drive
- UPCHP-298Extender tool
- UPCHP-299Extender tool
- UPCHP-399Extender tool
- UPCHP-297Extender tool
- UPCHP-292Extender tool, with 1/4" hex drive
- UPCHP-277Extender tool
- UPCHN-290Extender tool
- UPCHP-278Extender tool, with 1/4" hex drive