ContactUsLinkComponent
LB Medium Large Capsules, MP Biomedicals
LB Medium Large Capsules, MP Biomedicals
Supplier:  MP Biomedicals
Certificates
undefined
LB Medium Large Capsules, MP Biomedicals
Supplier:  MP Biomedicals
Specifications
  • Storage temperature:
    Store sterile 2X YT Broth at ambient temperature (15...30 °C). Powder medium is hygroscopic, therefore minimize exposure to air to prolong shelf life of media.
Specifications