Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tongs
Crucible tongs, stainless steel, double curved
Crucible tongs, stainless steel, double curved
Crucible tongs, stainless steel, double curved
Catalog # SCNG426
Supplier:
SCHNEIDER GERD
Expand image
Crucible tongs, stainless steel, double curved
Catalog # SCNG426
Supplier:
SCHNEIDER GERD
Specifications
Length:
600 mm
Pk:
1
Type:
Double curved
Specifications
About this item
Product Details
Stainless steel 18/10.
Product Details