ContactUsLinkComponent
Spoon, powder, stainless steel
Spoon, powder, stainless steel
  SCNG2373
Spoon, powder, stainless steel
  SCNG2373
  • Length:
    170 mm
  • Material:
    Stainless steel 1.4301
  • Pk:
    1
  • Spoon size:
    42×10 mm
With long handle and longitudinal bowl shape.