Order Entry
Germany
Orders LinkContactUsLinkComponent
Cleaning needles

Cleaning needles

Cleaning needle, stainless steel, 45° tip, with connector for 6,5 mm hose, 19, 1550 mm
Catalog # STAN14050-0
Supplier:   STANHOPE SETA
Cleaning needles
Catalog # STAN14050-0
Supplier:  STANHOPE SETA
Cleaning needles

Cleaning needle, stainless steel, 45° tip, with connector for 6,5 mm hose, 19, 1550 mm

Price
Inquire For Price
Log in for availability
1 * 1 Pack.
Product Details & Documents

Stainless steel cleaning needles for use with the Seta Twin-Column Adsorption Apparatus.

Specifications
  • Gauge:
    19
  • Description:
    Cleaning needle, stainless steel, 45° tip, with connector for 6,5 mm hose
  • Length:
    1550 mm
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.