Orders LinkContactUsLinkComponent
Suction filters, BOLA
Suction filters, BOLA
Supplier:   Bohlender
Suction filters, BOLA
Supplier:  Bohlender
Certificates
Suction filters, BOLA
Suction filters, BOLA
4 Options Available Below
Product Details & Documents
These PTFE suction filters consist of a receiver with a female thread and an easily exchangeable frit. They are ideal prefilters in front of pump systems for protecting gaskets, pistons or column packings against premature wearing due to contaminations.
Good chemical resistance
Temperature resistant from −200 to +250 °C
Product Family Options
Product Information
  • Connector
    Material
    Pore size
    For tube I-Ø
    Ø
    Length
    Availability
    Price
  • BOHLF765-08
    Suction filter
    Suction filter
    UNF 1/4" 28G
    PTFE
    2 μm
    0,8 mm
    14 mm
    25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 96,00 €
    96,00 €
    1 * 1 ST
       96,00 €
  • BOHLF765-16
    Suction filter
    Suction filter
    UNF 1/4" 28G
    PTFE
    2 μm
    1,6 mm
    14 mm
    25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 104,00 €
    104,00 €
    1 * 1 ST
       104,00 €
  • BOHLF765-48
    Suction filter
    Suction filter
    UNF 1/4" 28G
    PTFE
    10 μm
    0,8 mm
    14 mm
    25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 67,40 €
    67,40 €
    1 * 1 ST
       67,40 €
  • BOHLF765-56
    Suction filter
    Suction filter
    UNF 1/4" 28G
    PTFE
    10 μm
    1,6 mm
    14 mm
    25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 71,00 €
    71,00 €
    1 * 1 ST
       71,00 €