ContactUsLinkComponent
Human pulmonary fibroblasts
Human pulmonary fibroblasts
Catalog # C-14035
Supplier:  PROMOCELL
CAS Number:  
undefined
Human pulmonary fibroblasts
Catalog # C-14035
Supplier:  PROMOCELL
CAS Number:  
Specifications
  • Cell Format:
    Cryopreserved
  • Pk:
    2 mL
  • Description:
    Human pulmonary fibroblasts (HPF), 1×10⁶ cells, dissolved in 200 μl RNAlater®
  • Seq:
    02
Specifications
About this item
Primary Human Pulmonary Fibroblasts (HPF) are isolated from human lung tissue
  • CD90 positive
  • Cryopreserved at second passage
  • Available from different tissues
This product can be used in the following applications. Wound healing research, Tissue engineering research, Regeneration studies, Toxicological research, Oncology, Epithelial-mesenchymal interaction studies.
They are often used for studying lung tissue repair, tissue remodeling after injury, or the response after pulmonary inflammation. Fibroblast disorders like pulmonary fibrosis or hypoxia-induced pulmonary hypertension can also be investigated with cultured HPF.
Ordering information: Human Pulmonary Fibroblasts can be ordered as cryopreserved or proliferating cells. Additional cell pellets stored in RNAlater® are available.