ContactUsLinkComponent
Human Uterine fibroblasts (HUF)
Human Uterine fibroblasts (HUF)
Catalog # C-14038
Supplier:  PROMOCELL
CAS Number:  
undefined
Human Uterine fibroblasts (HUF)
Catalog # C-14038
Supplier:  PROMOCELL
CAS Number:  
Specifications
  • Cell Format:
    Pelleted
  • Pk:
    2 mL
  • Description:
    Primary human uterine fibroblasts (HUF) pellet, 1×106 cells in RNAlater®
Specifications
About this item
Primary human uterine fibroblasts (HUF) are isolated from the uterine wall.
During pregnancy, the uterine wall undergoes deep but reversible structural and biochemical changes. The uterine wall is the target of hormones and growth factors that act simultaneously and lead to extensive modifications of the extracellular matrix. This and other morphological and biochemical effects can be studied in vitro using HUF.