Orders LinkContactUsLinkComponent
Penetration probes, K-Type
Penetration probes, K-Type
Supplier:   Greisinger
Penetration probes, K-Type
Supplier:  Greisinger
Certificates
Penetration probes, K-Type
Penetration probes, K-Type
3 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • Range
    Availability
    Price
  • GRSG600431
    Spring-loaded V4A tube Ø 3 mm with slim insertion tip, with plastic handle, anti-buckling glanding,1 m highly flexible silicone cable and 3,5 mm gold plated jack connector
    Response time (T90): water 0,4 m/s < 2 s, air 2 m/s ~40 s
    Spring-loaded V4A tube Ø 3 mm with slim insertion tip, with plastic handle, anti-buckling glanding,1 m highly flexible silicone cable and 3,5 mm gold plated jack connector
    Response time (T90): water 0,4 m/s < 2 s, air 2 m/s ~40 s
    −70…+200 °C
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • GRSG600514
    Spring-loaded V4A tube with Ø 1,5 mm slim insertion tip, with plastic handle, anti-buckling glanding, 1 m silicone cable and miniature flat plug
    Response time (T90): water 0,4 m/s ~3 s
    Spring-loaded V4A tube with Ø 1,5 mm slim insertion tip, with plastic handle, anti-buckling glanding, 1 m silicone cable and miniature flat plug
    Response time (T90): water 0,4 m/s ~3 s
    −65…+550 °C
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • 620-4708
    Spring-loaded V4A tube with Ø 3 mm needle-shaped insertion tip, with plastic handle, anti-buckling glanding, 1 m silicone cable and miniature flat plug
    Response time (T90): water 0,4 m/s ~5 s
    Spring-loaded V4A tube with Ø 3 mm needle-shaped insertion tip, with plastic handle, anti-buckling glanding, 1 m silicone cable and miniature flat plug
    Response time (T90): water 0,4 m/s ~5 s
    −65…+550 °C
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST