Orders LinkContactUsLinkComponent
Human osteoblasts
Human osteoblasts
Human osteoblasts (HOB-p), 1×106 cells, dissolved in 200 μl RNAlater®, Pelleted, 2 mL
Catalog # C-14071
Marketsource
Supplier:   PROMOCELL
Human osteoblasts
Catalog # C-14071
Supplier:  PROMOCELL
Marketsource
CAS Number:  
Human osteoblasts
Human osteoblasts
Human osteoblasts (HOB-p), 1×106 cells, dissolved in 200 μl RNAlater®, Pelleted, 2 mL
CAS Number:  
Price
250,00 €
Log in for availability
1 * 2 mL
Product Details & Documents
Primary human osteoblasts (HOB) are isolated from femoral trabecular bone tissue from the knee or hip joint region. They stain positive for alkaline phosphatase and can be mineralised using mineralisation medium.
Osteoblasts are of mesenchymal origin and are mainly responsible for bone formation and remodeling. In vivo, they produce a collagen type I rich extracellular matrix called the osteoid. During their development, osteoblasts undergo a maturation process called osteogenesis. When they become encased in the bone matrix, they stop proliferating and become osteocytes.
Delivery information: Primary Human Osteoblasts can be ordered as cryopreserved or proliferating cells. Cell pellets stored in RNAlater® are also available.
Specifications
Specifications
  • Cell Format:
    Pelleted
  • Pk:
    2 mL
  • Description:
    Human osteoblasts (HOB-p), 1×10<sup>6</sup> cells, dissolved in 200 μl RNAlater®
Specifications