Orders LinkContactUsLinkComponent
Human preadipocytes
Human preadipocytes
Marketsource
Supplier:   PROMOCELL
Human preadipocytes
Supplier:  PROMOCELL
Marketsource
Certificates
Human preadipocytes
Human preadipocytes
2 Options Available Below
Product Details & Documents
Primary Human White Preadipocytes (HWP) are isolated from adult subcutaneous or visceral adipose tissue from different locations (lot specific source information is available on request). Adipose tissue is crucial in energy storage and metabolic homeostasis. An increase in adipose tissue results either from an enlargement of mature adipocytes or from the differentiation of adipocyte precursor cells (preadipocytes) into new mature adipocytes. These preadipocytes are already present in adipose tissues. They can proliferate throughout adult life and replace the cells that have differentiated into mature adipocytes.
Since this differentiation process is controlled by a variety of growth factors and hormones, preadipocytes are suitable for the investigation of the physiological mechanisms controlling proliferation, differentiation, and function of adipose tissue.
The PromoCell adipocyte cell culture system provides preadipocytes with optimized growth media and serum-free differentiation media.
Ordering information: Primary Human White Preadipocytes can be ordered as cryopreserved or proliferating cells. Additional cell pellets stored in RNAlater® are available.
Product Family Options
Product Information
  • Cell Format
    Pk
    Availability
    Price
  • C-14072
    Human White Preadipocytes (HWP) subcutaneous, 1×106 cells, dissolved in 200 μl RNAlater®
    Human White Preadipocytes (HWP) subcutaneous, 1×106 cells, dissolved in 200 μl RNAlater®
    Pelleted
    2 mL
    Log in for availability
    Log in for availability
    1 * 2 mL
    1 * 2 mL - 258,00 €
    258,00 €
    1 * 2 mL
       258,00 €
    Specifications:
    Marketsource
    Marketsource
  • C-14073
    Human White Preadipocytes (HWP) visceral,, 1×106 cells, dissolved in 200 μl RNAlater®
    Human White Preadipocytes (HWP) visceral,, 1×106 cells, dissolved in 200 μl RNAlater®
    Pelleted
    2 mL
    Log in for availability
    Log in for availability
    1 * 2 mL
    1 * 2 mL - 258,00 €
    258,00 €
    1 * 2 mL
       258,00 €
    Specifications:
    Marketsource
    Marketsource