Order Entry
Germany
Orders LinkContactUsLinkComponent
Mononuclear cell medium

Mononuclear cell medium

Mononuclear Cell Medium (ready-to-use), 500 ml, 500 mL
Catalog # C-28030
Marketsource
Supplier:   PROMOCELL
Mononuclear cell medium
Catalog # C-28030
Supplier:  PROMOCELL
Marketsource
CAS Number:  
Mononuclear cell medium
Mononuclear cell medium

Mononuclear Cell Medium (ready-to-use), 500 ml, 500 mL

CAS Number:  
Price
301,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

PromoCell Mononuclear Cell Medium is a very rich short-term maintenance medium which provides utmost cell viability. It is suitable to maintain medium for up to 48 hours before proceeding with your experiments.

Recommended for human mononuclear cells from cord blood (hMNC-CB), human mononuclear cells from peripheral blood (hMNC-PB), CD14+ monocytes.

Delivery information: The medium consists of 1× 500 ml bottle of basal medium and 1× vial of SupplementMix.

Specifications

Specifications

  • Pk:
    500 mL
  • Description:
    Mononuclear Cell Medium (ready-to-use), 500 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.