ContactUsLinkComponent
Monocyte attachment medium
Monocyte attachment medium
Catalog # C-28051
Supplier:  PROMOCELL
CAS Number:  
undefined
Monocyte attachment medium
Catalog # C-28051
Supplier:  PROMOCELL
CAS Number:  
Specifications
  • Pk:
    250 mL
  • Description:
    Monocyte Attachment Medium (ready-to-use), 250 ml
Specifications
About this item
PromoCell Monocyte Attachment Medium allows for the efficient adherence selection of monocytes from freshly isolated mononuclear cells (MNC) while maintaining optimal cell health. Thus, the time consuming and costly immunomagnetic purification of monocytes is dispensable prior to experiments requiring adherent monocytes as a starting material.