ContactUsLinkComponent
Mesenchymal stem cell growth medium DXF
Mesenchymal stem cell growth medium DXF
Catalog # C-28019
Supplier:  PROMOCELL
CAS Number:  
undefined
Mesenchymal stem cell growth medium DXF
Catalog # C-28019
Supplier:  PROMOCELL
CAS Number:  
Specifications
  • Pk:
    500 mL
  • Description:
    Mesenchymal Stem Cell Growth Medium DXF (ready-to-use), 500 ml
Specifications
About this item
PromoCell Mesenchymal Stem Cell Growth Medium DXF is a chemically defined and xeno-free formula, which strongly supports self-renewal and maximum growth of multipotent MSC. It is devoid of all animal-derived components and substances of human origin with human serum albumin as the only exception. Fibronectin-coated plates are necessary in conjunction with this medium.