Mesenchymal stem cell differentiation medium
Marketsource
Supplier: PROMOCELL
Jump to:
About this item
PromoCell Mesenchymal Stem Cell Differentiation Media efficiently induce differentiation of MSC into adipogenic, chondrogenic, osteogenic or neurogenic lineages, respectively.
5 Options Available Below
Product Details & Documents
PromoCell Mesenchymal Stem Cell Differentiation Media efficiently induce differentiation of MSC into adipogenic, chondrogenic, osteogenic or neurogenic lineages, respectively.
Product Family Options
Product Information
- PkAvailabilityPrice
- C-28012Mesenchymal Stem Cell Chondrogenic Differentiation Medium (ready-to-use), 100 mlMesenchymal Stem Cell Chondrogenic Differentiation Medium (ready-to-use), 100 ml100 mLLog in for availability1 * 100 mL552,00 €Specifications:MarketsourceMarketsource
- C-28014Mesenchymal Stem Cell Chondrogenic Differentiation Medium, without inducers (ready-to-use), 100 mlMesenchymal Stem Cell Chondrogenic Differentiation Medium, without inducers (ready-to-use), 100 ml100 mLLog in for availability1 * 100 mL451,00 €Specifications:MarketsourceMarketsource
- C-28013Mesenchymal Stem Cell Osteogenic Differentiation Medium (ready-to-use), 100 mlMesenchymal Stem Cell Osteogenic Differentiation Medium (ready-to-use), 100 ml100 mLLog in for availability1 * 100 mL237,00 €Specifications:MarketsourceMarketsource
- C-28015Mesenchymal Stem Cell Neurogenic Differentiation Medium (ready-to-use), 100 mlMesenchymal Stem Cell Neurogenic Differentiation Medium (ready-to-use), 100 ml100 mLLog in for availability1 * 100 mL467,00 €Specifications:MarketsourceMarketsource
- C-28016Mesenchymal stem cell Adipogenic Differentiation Medium 2 (ready-to-use), 100 mlMesenchymal stem cell Adipogenic Differentiation Medium 2 (ready-to-use), 100 ml100 mLLog in for availability1 * 100 mL237,00 €Specifications:MarketsourceMarketsource