Preadipocyte differentiation medium
Preadipocyte Differentiation Medium Kit, 500 ml, Liquid, 500 mL
Catalog # C-27437
Marketsource
Supplier: PROMOCELL
Jump to:
Product Details & Documents
PromoCell Preadipocyte Differentiation Medium is a serum-free medium which induces the differentiation of preadipocytes into mature adipocytes.
Delivery information: The medium (ready-to-use) consists of 1× 500 ml bottle of basal medium and 1× vial of SupplementMix. Kit consists of 1× 500 ml bottle of basal medium and 1× SupplementPack (individual vials with pre-measured supplements).
Specifications
Specifications
- Media Format:Liquid
- Pk:500 mL
- Description:Preadipocyte Differentiation Medium Kit, 500 ml
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



