Orders LinkContactUsLinkComponent
Smooth muscle cell growth medium 2
Smooth muscle cell growth medium 2
Marketsource
Supplier:   PROMOCELL
Smooth muscle cell growth medium 2
Supplier:  PROMOCELL
Marketsource
Certificates
Smooth muscle cell growth medium 2
Smooth muscle cell growth medium 2
4 Options Available Below
Product Details & Documents
Smooth Muscle Cell Growth Medium 2 is a low-serum (5% V/V) medium designed to establish and maintain smooth muscle cell cultures.
The medium is optimised for primary human cells but can also be used for bovine, porcine, murine, dog and rat smooth muscle cells.
Delivery information: PromoCell Smooth Muscle Cell Growth Medium 3 is available as Medium (ready to use) or as Medium Kit. The Medium (ready to use) consists of 1× 500 ml bottle of basal medium and 1× vial of SupplementMix. The Medium Kit consists of a 500 ml bottle of Basal Medium and the SupplementPack (a set of individual vials with pre-measured supplements) allowing the user full control over the media formulation. Adding the SupplementMix or the SupplementPack to the Basal Medium results in the complete Growth Medium. Basal Medium (with or without phenol red) as well as the SupplementMix and SupplementPack can also be purchased separately.
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • C-22062
    Smooth Muscle Cell Growth Medium 2 (ready to use)
    Smooth Muscle Cell Growth Medium 2 (ready to use)
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 284,00 €
    284,00 €
    1 * 500 mL
       284,00 €
    Specifications:
    Marketsource
    Marketsource
  • C-22162
    Smooth Muscle Cell Growth Medium 2 Kit
    Smooth Muscle Cell Growth Medium 2 Kit
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 286,00 €
    286,00 €
    1 * 500 mL
       286,00 €
    Specifications:
    Marketsource
    Marketsource
  • C-22267
    Smooth Muscle Cell Basal Medium 2, phenol red-free
    Smooth Muscle Cell Basal Medium 2, phenol red-free
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 213,00 €
    213,00 €
    1 * 500 mL
       213,00 €
    Specifications:
    Marketsource
    Marketsource
  • C-22262
    Smooth Muscle Cell Basal Medium 2 (without supplements)
    Smooth Muscle Cell Basal Medium 2 (without supplements)
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 197,00 €
    197,00 €
    1 * 500 mL
       197,00 €
    Specifications:
    Marketsource
    Marketsource