Serological pipettes, Shorty, KIMBLE®
Catalog # KIML72115-25210
Supplier: KIMBLEKONTES
Specifications
- Colour code:Green
- Packed:1/bag; 100/inner pack
- Sterile:+
- Capacity:25 ml
- Seq:04
- Division:0,2/5,0 ml
- Pk:200
Specifications
About this item
Borosilicate glass, disposable, sterile. These pipettes are ideal for use in tissue culture work and other procedures performed under a fume hood or in other confined areas.
- 'Negative' graduations to increase the versatility of the pipette
- Calibrated To Deliver (TD)
- Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
Packaging: Wrapped in colour-coded paper/plastic packaging.