Cell culture flasks, KIMAX®, KIMBLE®
Catalog # KIML25615-500
Supplier: KIMBLEKONTES
Specifications
- Capacity:500 ml
- Seq:02
- Pk:6
Specifications
About this item
Cell culture flasks, manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
- With white ceramic enamel graduation
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Thick walled tubing neck with a plain fire polished finish sealed to an Erlenmeyer-shaped body. And the extended neck is designed to accept a metal cap, plastic closure, foam plug, or cotton plug for cell culture applications.