Polyvinyl alcohol, M.W. 115,000 ≥88%, EMPROVE® EXPERT Ph. Eur., USP, JPE suitable for use as excipient, hydrolysed, SAFC®
Supplier: Merck
Certificates
About this item
CAS Number:9002-89-5
:25213-24-5
Formula:[-CH₂CH(OH)-]n
MDL Number:MFCD00081922